Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sptf-1
Entrez Gene ID
172909
Gene Name
protein-coding
Gene Aliases
CELE_F45H11.1
Chromosome Location
Chr.I: 10366621 - 10371699 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026294.3 NP_001021465.1 4 - 5 742 61
NM_001373640.1 NP_001359696.1 4 - 5 742 61

View Full Product Details