Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
blmp-1
Entrez Gene ID
172917
Gene Name
protein-coding
Gene Aliases
CELE_F25D7.3
Chromosome Location
Chr.I: 10409302 - 10414773 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001264441.2 NP_001251370.1 2 - 3 326 98
NM_001264442.2 NP_001251371.1 2 - 3 326 98

View Full Product Details