Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sec-8
Entrez Gene ID
172924
Gene Name
Exocyst complex component 4
Gene Aliases
CELE_Y106G6H.7
Chromosome Location
Chr.I: 10446657 - 10471767 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060331.8 NP_492732.2 5 - 6 525 68

View Full Product Details