Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
duo-3
Entrez Gene ID
172925
Gene Name
protein-coding
Gene Aliases
CELE_Y106G6H.12
Chromosome Location
Chr.I: 10464289 - 10471056 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306534.2 NP_001293463.1 3 - 4 410 119

View Full Product Details