Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
imb-3
Entrez Gene ID
3565440
Gene Name
Importin N-terminal domain-containing protein
Gene Aliases
CELE_C53D5.6
Chromosome Location
Chr.I: 310795 - 314989 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058314.6 NP_490715.1 4 - 5 1912 61

View Full Product Details