Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-59
Entrez Gene ID
266819
Gene Name
Prion-like-(Q/N-rich)-domain-bearing protein
Gene Aliases
CELE_R119.4
Chromosome Location
Chr.I: 379069 - 381902 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058326.7 NP_490727.1 2 - 3 314 74

View Full Product Details