Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
acs-13
Entrez Gene ID
171643
Gene Name
protein-coding
Gene Aliases
CELE_Y65B4BL.5
Chromosome Location
Chr.I: 511044 - 519646 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306267.3 NP_001293196.1 2 - 3 282 80
NM_001379780.1 NP_001368382.1 1 - 2 132 80
NM_058343.5 NP_490744.1 2 - 3 264 80

View Full Product Details