Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-20
Entrez Gene ID
171678
Gene Name
protein-coding
Gene Aliases
CELE_F54A5.2
Chromosome Location
Chr.I: 963695 - 973133 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383432.1 4 - 5 563 62
NM_058397.7 NP_490798.1 9 - 10 1835 62

View Full Product Details