Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-20
Entrez Gene ID
171678
Gene Name
protein-coding
Gene Aliases
CELE_F54A5.2
Chromosome Location
Chr.I: 963695 - 973133 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058397.7 NP_490798.1 10 - 11 1918 65

View Full Product Details