Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-45
Entrez Gene ID
171689
Gene Name
protein-coding
Gene Aliases
CELE_ZK993.1
Chromosome Location
Chr.I: 1128324 - 1131765 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306270.3 NP_001293199.1 2 - 3 271 66
NM_058422.7 NP_490823.1 4 - 5 561 66

View Full Product Details