Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
haf-6
Entrez Gene ID
171694
Gene Name
protein-coding
Gene Aliases
CELE_Y48G8AL.11
Chromosome Location
Chr.I: 1162880 - 1175868 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306301.3 NP_001293230.1 6 - 7 1241 74
NM_058427.6 NP_490828.3 8 - 9 1809 74

View Full Product Details