Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
grh-1
Entrez Gene ID
190048
Gene Name
protein-coding
Gene Aliases
CELE_Y48G8AR.1
Chromosome Location
Chr.I: 1258199 - 1268155 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306482.4 NP_001293411.1 6 - 7 1387 77
NM_001306483.3 NP_001293412.1 2 - 3 268 77
NM_058436.5 NP_490837.2 6 - 7 1388 77
GenBank mRNA AY323527.1 - 6 - 7 1387 77

View Full Product Details