Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
spe-15
Entrez Gene ID
171712
Gene Name
protein-coding
Gene Aliases
CELE_F47G6.4
Chromosome Location
Chr.I: 1446497 - 1473654 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306501.3 NP_001293430.1 10 - 11 1488 69
NM_001306502.3 NP_001293431.1 6 - 7 954 69
NM_058455.5 NP_490856.1 17 - 18 3604 69

View Full Product Details