Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
acs-16
Entrez Gene ID
171714
Gene Name
AMP-binding domain-containing protein
Gene Aliases
CELE_F47G6.2
Chromosome Location
Chr.I: 1471075 - 1479923 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058458.7 NP_490859.1 7 - 8 1339 55

View Full Product Details