Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mppa-1
Entrez Gene ID
171737
Gene Name
protein-coding
Gene Aliases
CELE_Y71G12B.24
Chromosome Location
Chr.I: 1740641 - 1751062 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058487.7 NP_490888.2 2 - 3 734 81

View Full Product Details