Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tag-96
Entrez Gene ID
171755
Gene Name
protein-coding
Gene Aliases
CELE_M01D7.4
Chromosome Location
Chr.I: 1845200 - 1850523 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058508.4 NP_490909.2 1 - 2 121 67

View Full Product Details