Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
afd-1
Entrez Gene ID
171776
Gene Name
protein-coding
Gene Aliases
CELE_W03F11.6
Chromosome Location
Chr.I: 2160038 - 2209644 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026489.7 NP_001021660.2 8 - 9 1220 132
NM_001026490.6 NP_001021661.2 8 - 9 1218 132
NM_001306400.4 NP_001293329.1 8 - 9 1222 132
NM_001306401.3 NP_001293330.1 3 - 4 357 132
NM_001306402.3 NP_001293331.1 3 - 4 357 132
NM_001306403.3 NP_001293332.1 2 - 3 171 132
NM_001306404.3 NP_001293333.1 2 - 3 171 132
NM_001356800.4 NP_001343732.1 5 - 5 773 132
NM_058537.5 NP_490938.3 8 - 9 1214 132

View Full Product Details