Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lgc-51
Entrez Gene ID
184377
Gene Name
protein-coding
Gene Aliases
CELE_F12B6.3
Chromosome Location
Chr.I: 2242230 - 2247223 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306430.1 NP_001293359.1 1 - 2 140 91
NM_001306431.1 NP_001293360.1 1 - 2 140 91

View Full Product Details