Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
inx-19
Entrez Gene ID
171805
Gene Name
protein-coding
Gene Aliases
CELE_T16H5.1
Chromosome Location
Chr.I: 2451898 - 2464618 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381412.2 NP_001367775.1 3 - 4 726 63
NM_058582.7 NP_490983.2 5 - 6 919 63

View Full Product Details