Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
abtm-1
Entrez Gene ID
171807
Gene Name
protein-coding
Gene Aliases
CELE_Y74C10AR.3
Chromosome Location
Chr.I: 2472038 - 2485397 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026659.2 NP_001021830.1 4 - 5 485 97

View Full Product Details