Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
itx-1
Entrez Gene ID
171846
Gene Name
protein-coding
Gene Aliases
CELE_W03D8.6
Chromosome Location
Chr.I: 2802803 - 2815830 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001262315.4 NP_001249244.2 14 - 15 2139 93
NM_001262316.5 NP_001249245.2 14 - 15 2158 93
NM_001306453.2 NP_001293382.1 14 - 15 2158 93
NM_001306454.3 NP_001293383.1 11 - 12 1638 93
NM_001306455.3 NP_001293384.1 11 - 12 1638 93

View Full Product Details