Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dnj-28
Entrez Gene ID
171870
Gene Name
DNaJ domain (prokaryotic heat shock protein)
Gene Aliases
CELE_Y54E10BL.4
Chromosome Location
Chr.I: 2996401 - 3001382 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058683.4 NP_491084.1 4 - 5 1143 111

View Full Product Details