Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mod-5
Entrez Gene ID
171879
Gene Name
Transporter
Gene Aliases
CELE_Y54E10BR.7
Chromosome Location
Chr.I: 3034716 - 3047794 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058694.4 NP_491095.3 1 - 2 48 73

View Full Product Details