Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-64
Entrez Gene ID
171884
Gene Name
protein-coding
Gene Aliases
CELE_C45E1.1
Chromosome Location
Chr.I: 3094513 - 3109621 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025871.6 NP_001021042.2 5 - 6 614 113
NM_001025872.6 NP_001021043.2 3 - 4 508 113
NM_001356801.3 NP_001343721.1 5 - 5 596 113
GenBank mRNA AY204173.1 - 4 - 5 605 113
AY204174.1 - 3 - 4 530 113

View Full Product Details