Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rom-5
Entrez Gene ID
190260
Gene Name
Rhomboid domain-containing protein
Gene Aliases
CELE_Y54E10A.14
Chromosome Location
Chr.I: 3217296 - 3222955 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058724.6 NP_491125.4 5 - 6 749 98

View Full Product Details