Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cdt-1
Entrez Gene ID
171896
Gene Name
CDT1 domain-containing protein
Gene Aliases
CELE_Y54E10A.15
Chromosome Location
Chr.I: 3223160 - 3228104 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047242.4 NP_001040707.1 2 - 3 1168 117

View Full Product Details