Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mis-12
Entrez Gene ID
171927
Gene Name
protein-coding
Gene Aliases
CELE_Y47G6A.24
Chromosome Location
Chr.I: 3604513 - 3605744 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058783.8 NP_491184.1 5 - 6 386 71

View Full Product Details