Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ccb-1
Entrez Gene ID
171933
Gene Name
GuKc domain-containing protein
Gene Aliases
CELE_T28F2.5
Chromosome Location
Chr.I: 3641651 - 3647082 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058792.6 NP_491193.2 5 - 6 671 106

View Full Product Details