Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ftn-2
Entrez Gene ID
171934
Gene Name
Ferritin
Gene Aliases
CELE_D1037.3
Chromosome Location
Chr.I: 3680172 - 3681873 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058797.8 NP_491198.1 2 - 3 164 127

View Full Product Details