Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ric-19
Entrez Gene ID
191757
Gene Name
Resistance to inhibitors of cholinesterase protein 19
Gene Aliases
CELE_C32E8.7
Chromosome Location
Chr.I: 3778089 - 3781690 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058815.5 NP_491216.2 7 - 8 1383 70

View Full Product Details