Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inx-15
Entrez Gene ID
172004
Gene Name
Innexin
Gene Aliases
CELE_R12E2.9
Chromosome Location
Chr.I: 4159006 - 4161113 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058912.5 NP_491313.1 4 - 5 1017 98

View Full Product Details