Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inx-16
Entrez Gene ID
172005
Gene Name
Innexin-16
Gene Aliases
CELE_R12E2.5
Chromosome Location
Chr.I: 4162046 - 4165031 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058913.5 NP_491314.1 1 - 2 87 74

View Full Product Details