Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl-3
Entrez Gene ID
172032
Gene Name
protein-coding
Gene Aliases
CELE_Y47D9A.2
Chromosome Location
Chr.I: 4298285 - 4302245 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001348610.3 NP_001335530.1 2 - 3 558 100
NM_001382905.1 4 - 5 952 100

View Full Product Details