Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcy-28
Entrez Gene ID
172051
Gene Name
protein-coding
Gene Aliases
CELE_T01A4.1
Chromosome Location
Chr.I: 4481198 - 4508940 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026429.3 NP_001021600.1 15 - 16 3593 87
NM_001262699.2 NP_001249628.1 18 - 19 3487 87
NM_001372632.2 NP_001360115.1 18 - 19 3479 87
NM_001382937.1 14 - 15 3384 87

View Full Product Details