Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcy-28
Entrez Gene ID
172051
Gene Name
protein-coding
Gene Aliases
CELE_T01A4.1
Chromosome Location
Chr.I: 4481198 - 4508940 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026429.3 NP_001021600.1 3 - 4 1214 109
NM_001262699.2 NP_001249628.1 7 - 8 1144 109
NM_001372632.2 NP_001360115.1 7 - 8 1130 109
NM_001382937.1 3 - 4 1041 109

View Full Product Details