Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-52
Entrez Gene ID
183885
Gene Name
COLlagen
Gene Aliases
CELE_D1007.2
Chromosome Location
Chr.I: 4571192 - 4575038 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058993.3 NP_491394.2 4 - 5 851 70

View Full Product Details