Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpl-4
Entrez Gene ID
172074
Gene Name
60S ribosomal protein L4
Gene Aliases
CELE_B0041.4
Chromosome Location
Chr.I: 4645632 - 4646963 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059015.7 NP_491416.1 3 - 4 549 62

View Full Product Details