Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-26
Entrez Gene ID
172085
Gene Name
protein-coding
Gene Aliases
CELE_C10H11.6
Chromosome Location
Chr.I: 4721131 - 4723982 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128946.3 NP_001122418.1 4 - 5 658 78
NM_059034.7 NP_491435.2 4 - 5 719 78

View Full Product Details