Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
viln-1
Entrez Gene ID
172089
Gene Name
HP domain-containing protein
Gene Aliases
CELE_C10H11.1
Chromosome Location
Chr.I: 4745675 - 4759601 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059041.5 NP_491442.1 3 - 4 2228 79

View Full Product Details