Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
htp-3
Entrez Gene ID
172099
Gene Name
HORMA domain-containing protein
Gene Aliases
CELE_F57C9.5
Chromosome Location
Chr.I: 4827072 - 4829796 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059057.6 NP_491458.1 8 - 9 1236 81

View Full Product Details