Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
imb-1
Entrez Gene ID
172110
Gene Name
Importin N-terminal domain-containing protein
Gene Aliases
CELE_F28B3.8
Chromosome Location
Chr.I: 4920683 - 4926486 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059076.7 NP_491477.2 3 - 4 351 111

View Full Product Details