Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dyf-1
Entrez Gene ID
172122
Gene Name
protein-coding
Gene Aliases
CELE_F54C1.5
Chromosome Location
Chr.I: 4989548 - 4994675 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059093.7 NP_491494.2 1 - 2 145 114

View Full Product Details