Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pat-10
Entrez Gene ID
172127
Gene Name
Paralysed Arrest at Two-fold
Gene Aliases
CELE_F54C1.7
Chromosome Location
Chr.I: 5018783 - 5020358 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059100.7 NP_491501.1 4 - 5 614 68

View Full Product Details