Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- chp-1
- Entrez Gene ID
-
172138
- Gene Name
- CHORD Protein
- Gene Aliases
-
CELE_Y110A7A.13
- Chromosome Location
-
Chr.I: 5103581 - 5106049 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059118.6 |
NP_491519.1 |
4 - 5 |
1741 |
73 |
GenBank mRNA |
AF192264.1 |
- |
3 - 4 |
650 |
73 |
- Gene Symbol
- pas-3
- Entrez Gene ID
-
172139
- Gene Name
- Proteasome subunit alpha type-4
- Gene Aliases
-
CELE_Y110A7A.14
- Chromosome Location
-
Chr.I: 5103612 - 5106048 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383045.1 |
|
4 - 5 |
1744 |
73 |
- Gene Symbol
- spe-5
- Entrez Gene ID
-
172137
- Gene Name
- Vacuolar proton pump subunit B
- Gene Aliases
-
CELE_Y110A7A.12
- Chromosome Location
-
Chr.I: 5101665 - 5104913 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059117.3 |
NP_491518.1 |
3 - 4 |
651 |
73 |
View Full Product Details