Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
elpc-1
Entrez Gene ID
172143
Gene Name
Elongator complex protein 1
Gene Aliases
CELE_Y110A7A.16
Chromosome Location
Chr.I: 5113989 - 5118852 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377580.1 NP_001364720.1 5 - 6 653 116

View Full Product Details