Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mat-1
Entrez Gene ID
172145
Gene Name
protein-coding
Gene Aliases
CELE_Y110A7A.17
Chromosome Location
Chr.I: 5123904 - 5131036 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026543.7 NP_001021714.1 7 - 8 1164 74
NM_001377581.3 NP_001364721.1 6 - 7 1081 74

View Full Product Details