Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mtm-1
Entrez Gene ID
172148
Gene Name
Myotubularin-related protein 1
Gene Aliases
CELE_Y110A7A.5
Chromosome Location
Chr.I: 5134709 - 5137971 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059130.5 NP_491531.2 5 - 6 666 110

View Full Product Details