Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hcp-6
Entrez Gene ID
266830
Gene Name
protein-coding
Gene Aliases
CELE_Y110A7A.1
Chromosome Location
Chr.I: 5165148 - 5171316 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059136.4 NP_491537.1 1 - 2 190 67
NM_059137.5 NP_491538.1 1 - 2 186 67

View Full Product Details