Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pde-5
Entrez Gene ID
183125
Gene Name
Probable 3',5'-cyclic phosphodiesterase pde-5
Gene Aliases
CELE_C32E12.2
Chromosome Location
Chr.I: 5196073 - 5200263 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059143.4 NP_491544.3 12 - 13 1814 67

View Full Product Details