Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnj-4
Entrez Gene ID
172173
Gene Name
J domain-containing protein
Gene Aliases
CELE_C01G8.4
Chromosome Location
Chr.I: 5282083 - 5283941 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059157.7 NP_491558.1 6 - 7 633 122

View Full Product Details